missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ TT-2 TableTop Anesthesia System built in Dual Diverter Valve System

Catalog No. 04777100
Change view
Click to view available options
For Use With (Application):
Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04777100 Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777100 Supplier SOMNI Scientific™ Supplier No. TT17000

Allows the user to provide anesthesia through one or both diverter valves without disconnecting supply lines

  • Dual diverters built in, use one or both
  • Two 6 ft. color-coded supply tubing included
  • Can be customized with additional accessories and is extremely easy to setup
  • Compact and portable
  • Compatible with all vaporizer mounts
  • Machined from durable, chemical resistant anodized aluminum

Specifications

For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) VS-6002, VS-6001,VS-6004, VS-6006, VS-6005, VS-6008, OC-21003, RE-20101, WG-15003, MW-17005, IA-3009, IP-3004, IR-3006, EM-16001, WG-15001, WG-15002, NC-19900
Material Anodized Aluminum
Length (Metric) 30.48 cm
Width (Metric) 19.7 cm
Includes Vaporizer endcaps and two 6 ft. color coded fresh gas delivery lines with connectors

For Veterinary Use Only