Learn More
Thermo Scientific™ TSX Universal Series Ultra-Low Freezer package with Racks, Boxes, and LN2 Back-up System

The new TSX Universal Series ULT freezers herald a new era in performance, delivering an expanded setpoint range, tighter control, and faster recovery time. Coupled with universal voltage, whisper-quiet operation, and ENERGY STAR certification, TSX Universal seamlessly adapts to your workflow.
Supplier: Thermo Scientific™ TSX60086FARLN
Description
TSX Universal Series ULTs represent our most robust design to date, incorporating insights from over 80 years of experience to enhance every aspect of the system. From sealed system and cabinet construction to software architecture and electrical components, each element of the TSX Universal ULT platform has undergone significant testing and improvements.
Enhancements to our V-Drive cooling technology: By updating our variable speed compressors, enhancing our intelligent algorithms, and optimizing hardware with our new Universal V-Drive cooling technology, users can now benefit from universally better performance than the previous generation TSX Series ULT platform.
Adaptable across workflows: Thanks to our new and innovative Universal V-Drive technology, users will experience tighter performance with faster pull-down and recovery times. From high-use academic labs to longer-term storage facilities, you can now enjoy the convenience of using the same unit throughout your diverse workflows, streamlining processes and enhancing overall manageability.
Greener by design: In 2022, we elevated our Design for Sustainability(DfS) approach, positioning our R&D organization to expand, accelerate, and standardize how environmental considerations are embedded into product development. Our strategy focuses on five key impact areas: less hazardous, less waste, more energy efficient, responsibly packaged, and extended life.
ENERGY STAR™-certified: Built with an unwavering commitment to sustainability, the next-generation TSX Universal ULTs are ENERGY STAR certified, with our largest size using 43% less energy than the allowed amount.
Transparency in sustainability: We believe it should be easy to make informed purchasing decisions when looking for more sustainable products. By participating in the ACT label program, run by the non-profit organization My Green Lab, we can provide easy-to-use information about our ULTs' environmental impact.
Thermo Fisher Connect Platform: Designed with labs of the future in mind, the TSX Universal ULT freezers come with 5 V and Power-over-Ethernet (PoE) connections, enabling users to introduce connectivity solutions to help monitor, share, and analyze data, all of which help streamline processes and improve research outcomes.
Connect Platform features:
- Real-time instrument health status
- Customizable report downloads
- Mobile and web applications
- Notifications: in-app alerts, email and push messaging
- Measurement data: temperature, system data, settings
- Bulk download and expert capability across multiple units
- Alarms: system, environmental, connectivity
- Event notifications: warnings, user based events, door open
- Detailed event descriptions
- Up to 6-month notification card of active alarms/events
- Full (lifetime) historical data storage and report retrieval
- Summary health report generation (on demand and planned)
Long-term reliability testing: We have implemented a new long-term reliability testing protocol for our TSX Universal ULT freezers platform. This protocol involves rigorous and continuous testing of our ULTs in field and laboratory environments to simulate real world usage across application types.
Designed for reliability- Compressor, electronics, cabinet, and sealed system
- High usage test
- 20 parameter test procedure
- Vibration testing
Every unit tested and documented: 100% of TSX Universal ULT units undergo a comprehensive end-of-line testing process to ensure they meet our rigorous quality standards. We document the results in a factory acceptance test report and a certificate of calibration, which are provided with every unit produced.
Tested for reliability with Multi-ambient testing
- Tested at 15°C, 20°C and 32°C
- Minimum 16 open air temperature probes
- Peak variation, stability, uniformity
- Door open recovery
Industry-leading warranty: We confidently stand behind our products, and we take pride in offering an industry-leading warranty, which sets us apart from any other ULT supplier in the market.
Tested to help ensure quality-100% end of line test
- Temperature pull down
- Monitoring compressor cycling
- Defrost temperatures
- Warmup time
- Bottom out temperature
Chemical Identifiers
| -40°C to -86°C | |
| 1 | |
| Stainless Steel | |
| ENERGYSTAR, EPA SNAP, cULus, CE | |
| Audible and Visual High and Low Temperature Alarms, Wireless | |
| 100/230 V | |
| V-drive |
| Solid, Hinged | |
| Cold-rolled Steel | |
| 3 Adjustable, 1 Fixed | |
| Upright | |
| NEMA 5-15 | |
| 100/230 V, 50/60 Hz |
Specifications
| 28.8 cu. ft. | |
| -40°C to -86°C | |
| 1 | |
| Stainless Steel | |
| ENERGYSTAR, EPA SNAP, cULus, CE | |
| Audible and Visual High and Low Temperature Alarms, Wireless | |
| 100/230 V | |
| V-drive |
| 816 L | |
| Solid, Hinged | |
| Cold-rolled Steel | |
| 3 Adjustable, 1 Fixed | |
| Upright | |
| NEMA 5-15 | |
| 100/230 V, 50/60 Hz | |
| 600 Boxes |