missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Universal Series Ultra-Low Freezer package with Racks, Boxes, and LN2 Back-up System
GREENER_CHOICE

Catalog No. TSX40086FLN
Change view
Click to view available options
Holds:
400 Boxes
500 Boxes
600 Boxes
700 Boxes
Capacity (Metric):
549 L
682 L
816 L
949 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Capacity (Metric)
TSX40086FLN 400 Boxes 549 L
TSX50086FLN 500 Boxes 682 L
TSX60086FLN 600 Boxes 816 L
TSX70086FLN 700 Boxes 949 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. TSX40086FLN Supplier Thermo Scientific™ Supplier No. TSX40086FARLN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The new TSX Universal Series ULT freezers herald a new era in performance, delivering an expanded setpoint range, tighter control, and faster recovery time. Coupled with universal voltage, whisper-quiet operation, and ENERGY STAR certification, TSX Universal seamlessly adapts to your workflow.

TSX Universal Series ULTs represent our most robust design to date, incorporating insights from over 80 years of experience to enhance every aspect of the system. From sealed system and cabinet construction to software architecture and electrical components, each element of the TSX Universal ULT platform has undergone significant testing and improvements.

Enhancements to our V-Drive cooling technology: By updating our variable speed compressors, enhancing our intelligent algorithms, and optimizing hardware with our new Universal V-Drive cooling technology, users can now benefit from universally better performance than the previous generation TSX Series ULT platform.

Adaptable across workflows: Thanks to our new and innovative Universal V-Drive technology, users will experience tighter performance with faster pull-down and recovery times. From high-use academic labs to longer-term storage facilities, you can now enjoy the convenience of using the same unit throughout your diverse workflows, streamlining processes and enhancing overall manageability.

Greener by design: In 2022, we elevated our Design for Sustainability(DfS) approach, positioning our R&D organization to expand, accelerate, and standardize how environmental considerations are embedded into product development. Our strategy focuses on five key impact areas: less hazardous, less waste, more energy efficient, responsibly packaged, and extended life.

ENERGY STAR™-certified: Built with an unwavering commitment to sustainability, the next-generation TSX Universal ULTs are ENERGY STAR certified, with our largest size using 43% less energy than the allowed amount.

Transparency in sustainability: We believe it should be easy to make informed purchasing decisions when looking for more sustainable products. By participating in the ACT label program, run by the non-profit organization My Green Lab, we can provide easy-to-use information about our ULTs' environmental impact.

Thermo Fisher Connect Platform: Designed with labs of the future in mind, the TSX Universal ULT freezers come with 5 V and Power-over-Ethernet (PoE) connections, enabling users to introduce connectivity solutions to help monitor, share, and analyze data, all of which help streamline processes and improve research outcomes.

Connect Platform features:

  • Real-time instrument health status
  • Customizable report downloads
  • Mobile and web applications
  • Notifications: in-app alerts, email and push messaging
  • Measurement data: temperature, system data, settings
  • Bulk download and expert capability across multiple units
  • Alarms: system, environmental, connectivity
  • Event notifications: warnings, user based events, door open
  • Detailed event descriptions
  • Up to 6-month notification card of active alarms/events
  • Full (lifetime) historical data storage and report retrieval
  • Summary health report generation (on demand and planned)

Long-term reliability testing: We have implemented a new long-term reliability testing protocol for our TSX Universal ULT freezers platform. This protocol involves rigorous and continuous testing of our ULTs in field and laboratory environments to simulate real world usage across application types.

Designed for reliability- Compressor, electronics, cabinet, and sealed system

  • High usage test
  • 20 parameter test procedure
  • Vibration testing

Every unit tested and documented: 100% of TSX Universal ULT units undergo a comprehensive end-of-line testing process to ensure they meet our rigorous quality standards. We document the results in a factory acceptance test report and a certificate of calibration, which are provided with every unit produced.

Tested for reliability with Multi-ambient testing

  • Tested at 15°C, 20°C and 32°C
  • Minimum 16 open air temperature probes
  • Peak variation, stability, uniformity
  • Door open recovery

Industry-leading warranty: We confidently stand behind our products, and we take pride in offering an industry-leading warranty, which sets us apart from any other ULT supplier in the market.

Tested to help ensure quality-100% end of line test

  • Temperature pull down
  • Monitoring compressor cycling
  • Defrost temperatures
  • Warmup time
  • Bottom out temperature

Specifications

Capacity (English) 19.4 cu. ft.
Capacity (Metric) 549 L
Temperature Range -40°C to -86°C
Door Style Solid, Hinged
No. of Doors 1
Cabinet Material Cold-rolled Steel
Chamber Material Stainless Steel
Shelves 3 Adjustable, 1 Fixed
Certifications/Compliance ENERGYSTAR, EPA SNAP, cULus, CE
Style Upright
Monitoring Options Audible and Visual High and Low Temperature Alarms, Wireless
Plug Type NEMA 5-15
Voltage 100/230 V
Electrical Requirements 100/230 V, 50/60 Hz
Compressor V-drive
Holds 400 Boxes
Show More Show Less