Learn More
Thermo Scientific™ TSX Series High-Performance -30°C Auto Defrost Freezers
Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers offer a sustainable choice for clinical and laboratory storage.
Supplier: Thermo Scientific™ TSX5030FD
Description
Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers are designed with features that support sample protection and sustainability objectives for the storage reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials such as primers, siRNA molecules and fetal bovine serum (FBS) requiring -30°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Certified to NSF/ANSI 456 Vaccine Storage* standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
*For NSF/ANSI 456 compliance, units must be ordered with a certificate of calibration (6916-2)
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 50% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerant and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Temperature range of -35° to -15°C, factory setpoint at -30°C
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
Specifications
| High-Performance Freezer | |
| 1447 L | |
| Double Solid | |
| 8 | |
| Microprocessor | |
| Warranty varies by country, please contact us for details | |
| 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm) | |
| 37.8 in. | |
| 96.0 cm | |
| 78.5 in. | |
| 199.4 cm | |
| 56.5 in. | |
| 143.5 cm | |
| Wireless | |
| Smart-Vue Compatible | |
| NEMA 6-15 | |
| 208-230V, 60Hz | |
| Energy efficient | |
| 8 |
| 51.1 cu. ft. | |
| -35° to -15°C (-30°C) | |
| 2 | |
| UL, cUL, ENERGYSTAR | |
| TSX Series | |
| Automatic | |
| 37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm) | |
| 28.5 in. | |
| 72.4 cm | |
| 58 in. | |
| 147.3 cm | |
| 52.5 in. | |
| 133.4 cm | |
| Optional | |
| 15 A (CB) | |
| 208/230 V | |
| 50/60 Hz | |
| R290 |
Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.