missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance -30°C Auto Defrost Freezers
GREENER_CHOICE

Catalog No. TSX3030FA
Change view
Click to view available options
Capacity (English):
11.5 cu. ft.
23 cu. ft.
29.2 cu. ft.
51.1 cu. ft.
Plug Type:
NEMA 5-15
NEMA 6-15
Electrical Requirements:
115V, 60Hz
208-230V, 50/60Hz
208-230V, 60Hz
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Plug Type Electrical Requirements
TSX3030FA 29.2 cu. ft. NEMA 5-15 115V, 60Hz
TSX2330FA 23 cu. ft. NEMA 5-15 115V, 60Hz
TSX5030FA 51.1 cu. ft. NEMA 5-15 115V, 60Hz
TSX5030FD 51.1 cu. ft. NEMA 6-15 208-230V, 60Hz
TSX1230FD 11.5 cu. ft. NEMA 6-15 208-230V, 60Hz
TSX1230FA 11.5 cu. ft. NEMA 5-15 115V, 60Hz
TSX3030FD 29.2 cu. ft. NEMA 6-15 208-230V, 60Hz
TSX2330FD 23 cu. ft. NEMA 6-15 208-230V, 50/60Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. TSX3030FA Supplier Thermo Scientific™ Supplier No. TSX3030FA
Get Thermo Scientific Accessories with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers offer a sustainable choice for clinical and laboratory storage.

Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers are designed with features that support sample protection and sustainability objectives for the storage reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials such as primers, siRNA molecules and fetal bovine serum (FBS) requiring -30°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Certified to NSF/ANSI 456 Vaccine Storage* standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

*For NSF/ANSI 456 compliance, units must be ordered with a certificate of calibration (6916-2)

  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Up to 50% less energy usage than conventional refrigerant models*
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP)
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Temperature range of -35° to -15°C, factory setpoint at -30°C
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Optional chart recorders available
  • Warranty varies by country, please contact us for details

Wireless Monitoring Solution

Thermo Scientific™ DeviceLink HUB™, our easy-to-install solution, works with the Thermo Fisher Cloud to monitor conditions inside your TSX Series Laboratory Refrigerators and Freezers.

DeviceLink HUB™ connects users to their equipment, allowing them to receive critical lab equipment data and equipment notifications on their mobile and web applications:

  • Display temperature
  • Ambient temperature
  • Alarm and alerts notification

DeviceLink HUB™ is available in the following regions:

  • United States and Canada
  • 28 members of the European Union
  • Iceland, Liechtenstein, Norway, and Switzerland

Specifications

Type High-Performance Freezer
Capacity (English) 29.2 cu. ft.
Capacity (Metric) 827 L
Temperature Range -15°C to -35°C
Temperature (Setpoint) -35° to -15°C (-30°C)
Door Style Solid
No. of Doors 1
Shelves 4
Certifications/Compliance UL, cUL, NSF 456 (When Purchased with 6916-2 Calibration Certificate), NSF 456 (When Purchased with 6916-2 Calibration Certificate)
Style Upright
Display Microprocessor
Product Line TSX Series
Defrost Automatic
Dimensions (DxWxH) Exterior 37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior 28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Dimensions (L x W x H) Exterior 37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Depth (English) Exterior 37.8 in.
Depth (English) Interior 28.5 in.
Depth (Metric) Exterior 96.0 cm
Depth (Metric) Interior 72.3 cm
Height (English) Exterior 78.5 in.
Height (English) Interior 58.0 in.
Height (Metric) Exterior 199.4 cm
Height (Metric) Interior 147.3 cm
Width (English) Exterior 34.0 in.
Width (English) Interior 30 in.
Width (Metric) Exterior 86.4 cm
Width (Metric) Interior 76.2 cm
Monitoring Options Wireless
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Amperage 15 A
Plug Type NEMA 5-15
Voltage 115 V
Electrical Requirements 115V, 60Hz
Frequency 60 Hz
Compressor V-drive
Refrigerant R290
Shipping Weight (English) 439 lb.
Shipping Weight (Metric) 199 kg
Weight (English) 371 lb.
Weight (Metric) 168 kg
Rack Capacity 4
Show More Show Less
What is the temperature range that can be maintained by TSX Series High-Performance -30 degrees C Auto Defrost Freezers?

TSX Series High-Performance -30 degrees C Auto Defrost Freezers can maintain temperatures between -35 and -15 degrees C with a setpoint of -30 degrees C.

Can the TSX Series High-Performance -30 degrees C Auto Defrost Freezers be locked?

Yes, The TSX Series freezers come with standard door locks to offer comfort and assurance, helping ensure the security of the stored materials.

Additionally, the units are equipped with a key-operated triple position switch for main power and alarm, which locks in temperature and alarm setpoints to minimize setpoint error and prevent tampering.

For enhanced access control, E-lock adapter kits are available. These E-locks provide access control with audit-trail capabilities and temperature monitoring. They don't require electricity and are available in both factory- and customer-installed options for various lock systems such as CompX™, Pyxis™, Omnicell™, and McKesson™locks. For additional details, refer to the TSX Series High-Performance Refrigerators and Freezers Brochure.

Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.