missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSX Series High-Performance Refrigerator, Solid Door, NEMA 5-15 PROMO
Thermo Scientific™ TSX Series high-performance laboratory refrigerators offer a sustainable choice for clinical and laboratory refrigeration.
Supplier: Thermo Scientific™ TSX2305SAPM
Description
- Designed to safely store medical and laboratory products, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2°C to 8°C
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 40% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerant and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Operating temperature range of 2° to 8°C, factory setpoint at 5°C
- Choice of glass or solid door models
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
Specifications
| TSX Series High-Performance Refrigerator, Solid Door | |
| 23 cu. ft. | |
| 3°C to 7°C (5°C) | |
| Solid | |
| 4 | |
| Microprocessor | |
| Automatic | |
| Optional | |
| 28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm) | |
| 37.8 in. | |
| 96.2 cm | |
| 78.6 in. | |
| 199.6 cm | |
| 28 in. | |
| 71.1 cm | |
| 15 A | |
| 115 V | |
| 60 Hz | |
| 426 lb. | |
| 361 lb. |
| High-Performance Refrigerator | |
| 650 L | |
| R290 | |
| 1 | |
| UL, ENERGYSTAR, cUL | |
| TSX Series | |
| Alarm, Recorder | |
| Smart-Vue Compatible | |
| 37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm) | |
| 28.5 in. | |
| 72.3 cm | |
| 58 in. | |
| 147.3 cm | |
| 24 in. | |
| 61 cm | |
| NEMA 5-15 | |
| 115 V, 60 Hz | |
| Energy efficient | |
| 193 kg | |
| 164 kg |