missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSX Series High-Performance Lab Refrigerator PROMO
Innovative refrigeration solutions for healthcare and research needs.
Supplier: Thermo Scientific™ TSX4505GADL
Description
- Designed to safely store medical and laboratory products, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2°C to 8°C
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 40% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerant and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Operating temperature range of 2° to 8°C, factory setpoint at 5°C
- Choice of glass or solid door models
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
Specifications
| High-Performance Refrigerator | |
| 1297 L | |
| R290 | |
| 8 Wire | |
| Microprocessor | |
| Warranty varies by country, please contact us for details | |
| Alarm, Recorder | |
| Smart-Vue Compatible | |
| 36.6 x 56.5 x 78.5 in (93.1 x 143.5 x 199.4 cm) | |
| 28.5 in. | |
| 72.4 cm | |
| 58 in. | |
| 147.3 cm | |
| 52.5 in. | |
| 133.4 cm | |
| NEMA 5-15 | |
| 115 V, 60 Hz | |
| Energy efficient | |
| 277 kg |
| 45.8 cu. ft. | |
| 3°C to 7°C (5°C) | |
| Sliding Glass | |
| UL, cUL, ENERGYSTAR | |
| TSX Series | |
| Automatic | |
| Optional | |
| 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm) | |
| 36.6 in. | |
| 93.1 cm | |
| 78.5 in. | |
| 199.4 cm | |
| 56.5 in. | |
| 143.5 cm | |
| 15 A (CB) | |
| 115 V | |
| 60 Hz | |
| 611 lb. |