missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSX Series High-Performance Refrigerator, Glass Door, NEMA 5-15 PROMO
Thermo Scientific™ TSX Series high-performance laboratory refrigerators offer a sustainable choice for clinical and laboratory refrigeration.
Supplier: Thermo Scientific™ TSX2305GAPM
Description
- Designed to safely store medical and laboratory products, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2°C to 8°C
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 40% less energy usage than conventional refrigerant models
- Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Operating temperature range of 2° to 8°C, factory setpoint at 5°C
- Choice of glass or solid door models
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
Specifications
| TSX Series High-Performance Refrigerator, Glass Door | |
| 23 cu. ft. | |
| 3°-7°C (5°C) | |
| Glass | |
| 4 | |
| Microprocessor | |
| Automatic | |
| Optional | |
| 28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm) | |
| 15 A | |
| 115 V | |
| Energy efficient | |
| 204 kg | |
| 175 kg |
| High-Performance Refrigerator | |
| 650 L | |
| R290 | |
| 1 | |
| UL | |
| TSX Series | |
| Alarm, Recorder | |
| Smart-Vue Compatible | |
| 37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm) | |
| NEMA 5-15 | |
| 60 Hz | |
| 450 lb. | |
| 385 lb. |