missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSV Value Undercounter Refrigerator

Catalog No. TSV05RPSA
Change view
Click to view available options
Type:
General Purpose ADA Refrigerator
General Purpose Refrigerator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
TSV05RPSA General Purpose Refrigerator
TSV05RPAA General Purpose ADA Refrigerator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TSV05RPSA Supplier Thermo Scientific™ Supplier No. TSV05RPSA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Undercounter Refrigerator

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Undercounter Refrigerator

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish

Specifications

Type General Purpose Refrigerator
Capacity (English) 5.5 cu. ft.
Capacity (Metric) 155.7 L
Temperature Range 1°C to 12°C
Temperature (Setpoint) Adjustable
Refrigerant R600a hydrocarbon refrigerant
Door Style Hinged
No. of Doors 1
Hinge Side Right
Shelves 3 Adjustable
Cabinet Material Cold-rolled Steel
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Style Upright
Casters 2 Adjustable Feet in Front
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime 10 years
Warranty 12 months
Defrost Manual
Min. Clearance 4 in. on all sides
Monitoring Options Remote Alarm Contacts
Access Security Key Lock
Compressor Hermetic
Dimensions (DxWxH) Exterior 23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1cm)
Dimensions (D x W x H) Interior 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm)
Amperage 0.8 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115V 60Hz
Frequency 60 Hz
Energy Usage .31 kWh/Day
Shipping Weight (English) 95 lb.
Shipping Weight (Metric) 43 kg
Show More Show Less