missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSV Value Lab Refrigerator/Freezer

Catalog No. TSV07CPSA
Change view
Click to view available options
Capacity (English) Refrigerator:
5.1 cu. ft.
7.1 cu. ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Refrigerator
TSV07CPSA 7.1 cu. ft.
TSV05CPSA 5.1 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TSV07CPSA Supplier Thermo Scientific™ Supplier No. TSV07CPSA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Refrigerator/Freezer.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish

Specifications

Type General Purpose Combo
Capacity (English) Refrigerator 7.1 cu. ft.
Capacity (Metric) Refrigerator 201 L
Temperature Range (Freezer) -12°C to -20°C
Temperature Range (Refrigerator) 1°C to 12°C
Door Style Hinged, Open Left to Right
No. of Doors 2
Drawers 1
Shelves 3 Adjustable (Refrigerator); 1 (Freezer)
Door Shelves 3
Compressor Hermetic
Defrost Automatic (Refrigerator), Manual (Freezer)
Access Security None
Refrigerant R600a hydrocarbon refrigerant
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Casters 2 adjustable feet in front
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor general storage of non-critical samples and reagents
Min. Clearance 12 in. on top; 4 in. sides and rear
Model TSV07CPSA
Typical Lifetime 10 years
Warranty 12 months
Dimensions (DxWxH) Exterior 21.75 x 21.5 x 56.63 in. (55.2 x 54.6 x 144 cm)
Dimensions (D x W x H) Interior Refrigerator: 17 x 17.5 x 36.5 in. (43.2 x 44.5 x 92.7 cm)Freezer: 16 x 15.5 x 10.38 in. (40.6 x 39.4 x 26.4 cm)
Energy Usage .39 kWh/Day
Amperage 1A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115V 60Hz
Frequency 60 Hz
Shipping Weight (Metric) 45 kg
Show More Show Less