missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSV Value Convertible Freezer/Refrigerator
Flexible and economical laboratory storage.
Supplier: Thermo Scientific™ TSV20FPSA
Description
Thermo Scientific TSV Value freezers and refrigerators offer economical and simple sample storage while also offering increased flexibility of operating as either a refrigerator or a freezer with the push of a button.
The TSV20FSPSA ships as an auto defrost freezer and the TSV20RPSA ships as a refrigerator, but both easily converts to a refrigerator or an auto defrost freezer respectively.
- Factory tested prior to shipment
- NRTL listed
- Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
- Adjustable temperature control
- Easy to clean, durable finish
Chemical Identifiers
| 21 cu. ft. | |
| 1°C to 12°C or -12°C to -24°C | |
| R600a | |
| 1 (1 compartment) | |
| 3 Adjustable | |
| White-enameled Steel | |
| 2 adjustable leveling feet | |
| Electronic control | |
| Automatic | |
| None | |
| Bottom-mounted | |
| 2.2A | |
| 115 V/60 Hz | |
| 244 lb. |
| 594 L | |
| Adjustable | |
| Hinged | |
| Right | |
| 4 fixed shelves | |
| Cyclopentane | |
| White | |
| Indoor general storage of non-critical samples and reagents | |
| 4 in. on sides, 4 in. at back, 11 in. on top | |
| 2 - refrigerator or freezer | |
| 28.94 x 32.76 x 76.77 in. (73.5 x 83.2 x 195 cm) | |
| NEMA 5-15 P | |
| 50 Hz | |
| 110.6 kg |
Specifications
| General Purpose Freezer (Convertible to Refrigerator) | |
| 594 L | |
| Adjustable | |
| Hinged | |
| Right | |
| 4 fixed shelves | |
| Cyclopentane | |
| 2 adjustable leveling feet | |
| Electronic control | |
| 10 Years | |
| Automatic | |
| 4 in. on sides, 4 in. at back, 11 in. on top | |
| 2 - refrigerator or freezer | |
| 22.5 x 26.5 x 69 in. (57.15 x 67.3 x 175.26 cm) | |
| 2.2A | |
| 115 V/60 Hz | |
| 244 lb. |
| 21 cu. ft. | |
| 1°C to 12°C or -12°C to -24°C | |
| R600a | |
| 1 (1 compartment) | |
| 3 Adjustable | |
| White-enameled Steel | |
| ABS plastic | |
| White | |
| Indoor general storage of non-critical samples and reagents | |
| 13 Months Parts and Labor | |
| Interior - Yes | |
| None | |
| Bottom-mounted | |
| 28.94 x 32.76 x 76.77 in. (73.5 x 83.2 x 195 cm) | |
| NEMA 5-15 P | |
| 50 Hz | |
| 110.6 kg |