missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSV Value Combo Refrigerator/Freezer

Catalog No. TSV18CPSA
Change view
Click to view available options
Capacity (Metric):
510 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
TSV18CPSA 510 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TSV18CPSA Supplier Thermo Scientific™ Supplier No. TSV18CPSA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value Refrigerator/Freezer.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, SNAP-compliant hydrocarbon refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto-defrost combination units

Specifications

Type Combination General Purpose Refrigerator/Freezer
Capacity (English) 18 cu. ft.
Capacity (Metric) 510 L
Temperature Range 1°C to 12°C
Temperature (Setpoint) Adjustable
Refrigerant R600a
Door Style Hinged
No. of Doors 2 (2 compartments)
Hinge Side Right
Shelves 2 Adjustable Shelves, 2 Drawers (Refrigerator); 1 Shelf (Freezer)
Door Shelves 3
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Casters 2 adjustable leveling feet
Color White
Control Type Electronic Controls
For Use With (Application) Indoor general storage of non-critical samples and reagents
Typical Lifetime 10 years
Defrost Automatic
Lighting Interior
Min. Clearance 4 in. on Sides, 4 in. at Back, 11 in. on Top
Compressor Bottom-mounted
Dimensions (D x W x H) Interior Refrigerator: 25.5 x 25 x 40 in. (64.7 x 63.5 x 101.6 cm) Freezer: 20.5 x 22.5 x 15.75 in. (52 x 57.1 x 40 cm)
Dimensions (L x W x H) Exterior 32.79 x 29.53 x 66.61 in. (83.3 x 75 x 169.2 cm)
Amperage 1.1A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V/60 Hz
Frequency 50 Hz
Weight (English) 224 lb.
Weight (Metric) 101.6 kg
Show More Show Less