missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSV Value Combo Refrigerator/Freezer PROMO
Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value Refrigerator/Freezer.
Supplier: Thermo Scientific™ TSV18CPSAPM
Description
- Factory tested prior to shipment
- NRTL listed
- Environmentally-friendly, SNAP-compliant hydrocarbon refrigerants
- Adjustable temperature control
- Easy to clean, durable finish
- Manual and auto-defrost combination units
Spécifications
| Combination General Purpose Refrigerator/Freezer | |
| 510 L | |
| Adjustable | |
| Hinged | |
| Right | |
| 3 | |
| Cyclopentane | |
| cULus | |
| White | |
| Indoor general storage of non-critical samples and reagents | |
| Automatic | |
| 4 in. on Sides, 4 in. at Back, 11 in. on Top | |
| Refrigerator: 64.7 x 63.5 x 101.6 cm (25.5 x 25 x 40 in.); Freezer: 52 x 57.1 x 40 cm (20.5 x 22.5 x 15.75 in.) | |
| 1.1 A | |
| 115 V | |
| 50 Hz | |
| 101.6 kg |
| 18 cu. ft. | |
| 1°C to 12°C | |
| R600a | |
| 2 (2 compartments) | |
| 2 Adjustable | |
| Enameled Steel | |
| ABS Plastic | |
| 2 adjustable leveling feet | |
| Electronic Controls | |
| 10 years | |
| Interior | |
| Bottom-mounted | |
| 83.3 x 75 x 169.2 cm (32.79 x 29.53 x 66.61 in.) | |
| NEMA 5-15 P | |
| 115 V, 60 Hz | |
| 224 lb. |