missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSV Series 72 cu.ft. Lab Refrigerator PROMO
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents.
Specifications
| Capacity (English) | 72 cu. ft. |
|---|---|
| Capacity (Metric) | 2038 L |
| Temperature Range | 4°C to 6°C |
| Temperature (Setpoint) | 5°C |
| Refrigerant | R290 |
| Catalog Number | Mfr. No. | Cabinet Material | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Cabinet Material | Price | Quantity | |||||
|
TSV72RPGAPM
|
Thermo Scientific™
TSV72RPGAPM |
Painted Steel |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
TSV72RSGAPM
|
Thermo Scientific™
TSV72RSGAPM |
Stainless Steel |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Three self-closing glass doors
- Remote alarm contacts
- High and low temperature alarms
- Digital controller with adjustable temperature setpoint
- Door ajar alarm
- Optional stainless steel interior
Specifications
| 72 cu. ft. | |
| 4°C to 6°C | |
| R290 | |
| 3 | |
| 100 lb. | |
| cULus | |
| Digital | |
| One rear access port | |
| Automatic | |
| Key Lock | |
| 75.7 x 32 x 81 in. (1924 x 813 x 2057 mm) | |
| NEMA 5-15 | |
| 730 lb. |
| 2038 L | |
| 5°C | |
| Glass Self-closing with Lock | |
| 12 Adjustable | |
| 45 kg | |
| Freestanding | |
| Yes, customer installed | |
| 2 Years Parts and Labor, 3 Additional Years Compressor | |
| SMART VUE Compatible, Remote Alarm Contacts | |
| Bottom-mounted | |
| 72 x 28.5 x 61.75 in. (1829 x 724 x 1569 mm) | |
| 120 V, 60 Hz | |
| 318 kg |