Learn More
SignalChem Biotech DRAK1 (STK17A), Active 500ug
Catalog No. 502573086
Encompass
Recombinant full-length human DRAK1 (STK17A) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573086 Supplier SignalChem Biotech Supplier No. S3310G500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human DRAK1 (STK17A) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.