missing translation for 'onlineSavingsMsg'
Learn More

Wanzl TSS Clear Basket Label Holder

Catalog No. 11222120
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
60 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11222120 24 in.
11222121 36 in.
11222122 48 in.
11222123 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11222120 Supplier Wanzl Supplier No. T.TLB24CL
Only null left

Clear Basket Label Holder for use with TSS Shelving System.

  • Add-on option for additional customization
  • Clear
  • Three ways to mount

Specifications

Width (English) 24 in.
Color Clear
For Use With (Application) Shelving
Includes Basket Label Holder
Material Plastic