missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl TSS Chrome Wire Shelf
Description
- Add-on option for additional customization
- Offered in our Chrome Plus (CH) finish
- The heavy nickel plate process features a hard chromium finish with special brighteners to enhance and increase the life of the shelving.
Specifications
Specifications
| Color | Chrome |
| Material | Steel |
| Dimensions (L x W) | 18 x 24 in. |
| For Use With (Application) | Shelving |
| Shelf Type | Wire Shelf |
| Width (English) | 24 in. |