Learn More
ACROBiosystems Inc Recombinant Protein;Human TSLP Protein;HEK293;1MG;TSP,TSLP,TSLP R
Catalog No. 501657901
Encompass
Recombinant Protein;Human TSLP Protein;1MG;Human TSLP (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 8 kDa and 14-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 g/mL (100 L/well) can bind Human TSLP (Cat. No. TSP-H52Hb) with a linear range of 1.6-13 ng/mL (QC tested).;Immobilized Human TSLP (Cat. No. TSP-H52Hb) at 1 g/mL (100 L/well) can bind Anti-TSLP, Human IgG2 with a linear range of 0.5-8 ng/mL (Routinely tested).
Catalog No. 501657901 Supplier ACROBiosystems Inc Supplier No. TSPH52HB1MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human TSLP Protein;1MG;Human TSLP (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 8 kDa and 14-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 g/mL (100 L/well) can bind Human TSLP (Cat. No. TSP-H52Hb) with a linear range of 1.6-13 ng/mL (QC tested).;Immobilized Human TSLP (Cat. No. TSP-H52Hb) at 1 g/mL (100 L/well) can bind Anti-TSLP, Human IgG2 with a linear range of 0.5-8 ng/mL (Routinely tested).