Recombinant Protein;1MG;Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # A0A2K5TXV0-1 (R127A, R130S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.9 kDa. The protein migrates as 23-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 g/mL (100 L/well) can bind Cynomolgus TSLP (R127A, R130S), His Tag (Cat. No. TSP-C52H4) with a linear range of 1-4 ng/mL (QC tested).;Immobilized Cynomolgus TSLP (R127A, R130S), His Tag (Cat. No. TSP-C52H4) at 5 g/mL (100 L/well) can bind Monoclonal Anti-Human TSLP Antibody, Human IgG2 with a linear range of 0.5-8 ng/mL (Routinely tested).