Learn More
Tosoh Bioscience Llc TSKgel SuperSW3000, Column, Gel Filtration Chromatography
Supplier: Tosoh Bioscience Gmbh TB21485
Column, Gel Filtration Chromatography; Tosoh TSKgel SuperSW3000; Media: Silica; Particle: 4 micrometer; 2.0mm x 30cm Length; Stainless Steel