missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ QAE-2SW Guardgel Kit, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021784
Change view
Click to view available options
pH Stability:
2 to 7.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Stability
111021784 2 to 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021784 Supplier Merck Emd Millipore Supplier No. 807646

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

0-100% organic solvent, spherical silica matrix

  • 5 mL Guardgel packing material
  • Column holder, consisting of external reducing union, cap, frit, 2 ferrules
  • Column, 25 x 9.5 mm OD x 6 mm ID, 2 μm frit at 10 mm depth, 1/16 in. exit
  • 2 Replacement frits
  • Capillary tubing, 5 cm x 1/16 in. OD x 0.01 in. ID
  • Nuts and ferrules, 10-32 threads for 1/16 in. tubing, 2 each
  • All hardware is 316 stainless steel
  • Shipped in methanol.

Specifications

Type Ammonium Ion, Quaternary Phase Matrix Active Group
Length (Metric) 2.5 cm
Material Sstainless steel
Description Uses seperation technique anion exchange
Diameter (Metric) Inner 6 mm
pH Stability 2 to 7.5
Size (Metric) 5 μm
Operating Temperature (Metric) 10°C to 45°C
For Use With (Equipment) L14 Per USP

TSKgel is a registered trademark of Tosoh Corporation