missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Lipopropak XL HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021964
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021964 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021964 Supplier Merck Emd Millipore Supplier No. 818557
Only null left

A specialty column for the analysis of lipoprotein classes in serum using a dedicated eluent. The particles in TSKgel Lipopropak XL columns are spherical, 10 μm porous polymethacrylate beads. The separation of the lipoproteins takes place by size exclusion.

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1 mL/min
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Shape Sphere
Particle Size 10 μm
pH 2 to 12
Solvents ≤ 20% Organic Solvent
Temperature 80°C
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Shipping Condition Shipped in TSKeluent LP-1 (Aqueous)
Matrix Hydroxylated Methacrylate Phase/Hydroxylated Methacrylic Polymer
Material Stainless Steel
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation