missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ GMHXL-L HPLC Column, 6 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021879
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021879 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021879 Supplier Merck Emd Millipore Supplier No. 816652
Only null left

TSKgel GMHXL-L Mixed-Bed Columns are packed with a mixture of rigid, polymer beads, varying in average pore size. Ideal for the analysis of organic-soluble industrial polymers.

  • Mixed bed, or linear columns are used when a broad range of polymer molecular weights is expected in the sample
  • This column gives high resolution of polymers with a molecular weight up to about 100,000 Da, although the exclusion limit is higher
  • THF shipping solvent can be replaced by benzene, chloroform, toluene and xylene, but not with dichloromethane and dichloroethane

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min
Length (Metric) 30 cm
Max. Pressure 35 bar
Particle Size 6 μm
Temperature 80°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation