missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ G2500PW HPLC Column, 17 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021874
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021874 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021874 Supplier Merck Emd Millipore Supplier No. 816248
Only null left

Use TSKgel G2500PW for the analysis of water-soluble polymers with molecular weights of less than 3,000 Da.

  • Shipped in water
  • Salt Concentration: ≤ 0.5 M

Specifications

Diameter (Metric) 21.5 mm
Flow Rate 8 mL/min
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Size 17 μm
pH 2 to 12
Pore Size <200 Å
Solvents ≤ 20% Organic Solvent
Temperature 80°C
USP Type L25, L38, L39
Stationary Phase Hydroxylated Methacrylate
Shipping Condition Shipped aq. soln.
Matrix Hydroxylated Methacrylate Phase/Polymer Particle Platform
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation