Learn More
Tosoh Bioscience Llc TSKGEL DNA-NPR 4.6X75 DFS Item
Supplier:  Tosoh Bioscience Gmbh TB18249
Column, Nonporous (NPR) HPLC; Anion Exchange; Tosoh TSKgel DNA-NPR; Media: Nonporous polymer; Particle: 2.5 micrometer; 4.6mm I.D. x 7.5cm Length
Catalog No. 50851482
Only null left
Explore available promotions