missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG Series Laboratory Freezers

Catalog No. TSG1225FA
Change view
Click to view available options
Capacity (English):
12 cu. ft.
23 cu ft.
50 cu.ft.
Dimensions (DxWxH) Exterior:
22 x 20 x 52 in.
29 x 53 x 58 in.
39 x 24 x 58 in.
No. of Doors:
1
2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (DxWxH) Exterior No. of Doors
TSG1225FA 12 cu. ft. 22 x 20 x 52 in. 1
TSG2325FA 23 cu ft. 39 x 24 x 58 in. 1
TSG5025FA 50 cu.ft. 29 x 53 x 58 in. 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. TSG1225FA Supplier Thermo Scientific™ Supplier No. TSG1225FA
Get Thermo Scientific Accessories with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TSG Auto-defrost Freezers offer tight temperature performance at -25°C setpoint with a 12-hour controller backup, remote alarm contacts and a 4-year parts and labor warranty. Available in single and double-door configurations, the freezers are sustainably made at a zero waste-to-landfill facility.

TSG Series Laboratory Freezers are built from the ground up to meet your cold storage needs, offering quiet operation, optimal performance, and sustainable design. The TSG Series is ready to go out of the box, complete with pre-installed casters, onboard alarms, a 12-hour controller backup system, and remote alarm contacts. These auto-defrost freezers also give you peace of mind knowing that your samples are secure with robust temperature performance, all backed by a 4-year parts and labor warranty.

Designed for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory grade storage requiring tight temperature performance
  • Non-invasive adaptive defrost with time and temperature guided cycle
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Easy to clean and adjust epoxy coated shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • E-Lock Adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules

Services and support

  • Industry Leading 60-month compressor warranty (varies by country, please contact us for details)
  • Optional Validation Testing Programs include field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs

Specifications

Capacity (English) 12 cu. ft.
Capacity (Metric) 400 L
Temperature Range -15°C to -29°C
Temperature Resolution 1°C
Temperature Sensor Yes
Temperature (Setpoint) -25°C
Door Style Solid Swing
No. of Doors 1
Cabinet Material Aluminum
Chamber Material Aluminum
Shelves 4
Capacity (English) Shelf 100 lb.
Capacity (Metric) Shelf 45 kg
Certifications/Compliance RoHS, REACH, cULus, EPA SNAP, EPA SNAP
Style Upright
Casters Yes
For Use With (Application) General Purpose
Warranty 4 Years (Parts and Labor)
Defrost Automatic
Dimensions (DxWxH) Exterior 22 x 20 x 52 in.
Monitoring Options Alarm Module, Battery Back-up
Access Security Yes
On-Board Datalogging Yes
Backup Systems Yes
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V/60 Hz
Frequency 60 Hz
Compressor Bottom-mounted
Refrigerant R290
Shipping Weight (English) 265 lb.
Shipping Weight (Metric) 120 kg
Show More Show Less