Learn More
Thermo Scientific™ TSG Series Solid Door Laboratory Refrigerators - Trade-Up Program
Modernize your solid door lab refrigerator through the available trade-up program and take advantage of significant savings and disposal options. The TSG Solid Door Refrigerators offer excellent performance for sample storage needing 2 to 8°C, and an outstanding 4-year parts and labor warranty.
Supplier: Thermo Scientific™ TSG3005SAPM
Description
The drive for sample integrity and security
- Designed to safely store media, reagents, and other laboratory-grade products requiring a temperature range of 3°C to 7°C
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Standard door locks provide comfort and assurance
Enabling compliance
- Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863
Designed for an ergonomic and comfortable experience
- Ergonomically designed door handle
- Convenient, easy to clean and adjustable epoxy coated full and half shelves
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Bright, LED interior lighting
- Self-closing multipaned solid doors with 90° stop to assist with inventory loads
Verified sustainability
- US EPA ENERGY STAR Certified
- ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
- Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
- Manufactured in an award-winning, zero-waste to landfill facility
Ready for connectivity and monitoring
- Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make it your own
- Extra and Alternate wire baskets, shelves, and drawers
- Stainless Steel interior finish available
- E-Lock Adapter kits
Specifications
| 29.2 cu. ft. | |
| 3°C to 7°C | |
| R290 | |
| 1 | |
| 100 lb. | |
| Painted Steel | |
| White (Painted) | |
| Capacitive Touch | |
| 4 Years Parts and Labor + 1 Year Compressor | |
| Key Lock | |
| 28.5 x 30.0 x 58.0 in. (723 x 762 x 1473 mm) | |
| 115 V, 60 Hz | |
| 208 kg |
| 827 L | |
| +5°C | |
| Solid | |
| 4 Adjustable | |
| 45 kg | |
| Painted | |
| cULus, ENERGYSTAR | |
| 1 Rear Access Port | |
| SmartVue Pro, Remote Alarm Contacts | |
| 37.9 x 34.0 x 78.5 in. (960 x 864 x 1994 mm) | |
| NEMA 5-15 | |
| 459 lb. |