missing translation for 'onlineSavingsMsg'
Learn More

Tryptophol, 97%

Catalog No. AAL0255503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0255503 1 g
AAL0255506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0255503 Supplier Thermo Scientific Chemicals Supplier No. L0255503

CAS: 526-55-6 | C10H11NO | 161.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant for preparation of inhibitors of the C-terminal domain of RNA polymerase II and their antitumor activities, anti-HIV-1 agents, inhibitors of Protein-Protein Interactions, partial agonists of the serotonin 5-HT1A receptor, growth hormone secretagogues, vascular endothelial growth factor (VEGF) inhibitors, A2B adenosine receptor ligands, potential detoxification inhibitors of the crucifer phytoalexin brassinin, inhibitors of interleukin and dual binding site acetyl cholinesterase inhibitors.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in water at 25°C 2749 mg/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005659
InChI Key MBBOMCVGYCRMEA-UHFFFAOYSA-N
Synonym tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
PubChem CID 10685
ChEBI CHEBI:17890
IUPAC Name 2-(1H-indol-3-yl)ethanol
SMILES OCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 56°C to 59°C
Boiling Point 174°C (2 mmHg)
Flash Point 255°C (491°F)
Quantity 1 g
Beilstein 125553
Merck Index 14,9798
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material Tryptophol
EINECSNumber 208-393-2
RTECSNumber KL3685000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only