missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Agar EP/USP/JP/BP (Dehydrated)

Catalog No. p-200112237
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0131B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXCM0131B Supplier Thermo Scientific™ Supplier No. CM0131B
Only null left

Grow a wide variety of microorganisms with this general purpose medium specified in international pharmacopoeia.

Grow a wide variety of microorganisms with Thermo Scientific ™ Oxoid ™ Tryptone Soya Agar (Dehydrated). A general purpose agar medium containing two peptones, Tryptone Soya Agar supports the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition1.

Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopoeia and food testing methods for the growth of a wide variety of organisms. The product is tested to the pharmacopoeia2, 3, 4, 5 and is mainly used as reference medium when testing selective media.

  • Recommended as a reference medium for testing selective media
  • Conforms to formulations detailed in the International pharmacopoeia

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description Tryptone Soya Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.5L medium

For Laboratory Use Only