Learn More
Thermo Scientific™ Tryptone Soy Broth (Soybean Casein Digest Medium EP/USP/BP/JP) (Dehydrated)
Grow a wide variety of microorganisms with this general purpose medium containing two peptones.
Supplier:  Thermo Scientific™ CM0129T
 View more versions of this product
Catalog No. OXCM0129T
Only null left
Explore available promotions
Description
Description
Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth EP/USP/BP/JP (Dehydrated). The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.
Use Tryptone Soya Broth to support the luxuriant growth of many fastidious organisms, without the addition of serum, while conforming to the formulations detailed in various International Pharmacopoeias.
  • Suitable for tube dilution method of antibiotic susceptibility testing
  • Recommended in the USP for the recovery of organisms after sterilization processes4
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Tryptone Soy Broth (Soybean Casein Digest Medium USP)
Dehydrated Culture Media
For 167.0L medium
Powder
5 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Laboratory Use Only