missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soy Broth (Soybean Casein Digest Medium EP/USP/BP/JP) (Dehydrated)

Catalog No. OXCM0129R
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0129R 2.5 kg
OXCM0129B 500 g
OXCM0129T 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OXCM0129R Supplier Thermo Scientific™ Supplier No. CM0129R
Only null left

Grow a wide variety of microorganisms with this general purpose medium containing two peptones.

Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth EP/USP/BP/JP (Dehydrated). The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Use Tryptone Soya Broth to support the luxuriant growth of many fastidious organisms, without the addition of serum, while conforming to the formulations detailed in various international pharmacopoeias.

  • Suitable for tube dilution method of antibiotic susceptibility testing
  • Recommended in the USP for the recovery of organisms after sterilization processes4

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications

Description Tryptone Soy Broth (Soybean Casein Digest Medium USP)
Form Powder
Product Type Dehydrated Culture Media
Quantity 2.5 kg
Yield For 83.5L medium

For Laboratory Use Only