missing translation for 'onlineSavingsMsg'
Learn More

Hardy Diagnostics™ Tryptic Soy Broth (TSB)

Catalog No. 23001828
Change view
Click to view available options
:
15 mL Fill
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23001828 15 mL Fill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23001828 Supplier Hardy Diagnostics™ Supplier No. K83
Only null left

Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi.

  • The U.S. Pharmacopeia National Formulary (USP) describes its use for sterility testing.
  • This product is not intended to be used for the diagnosis of human disease.

Specifications

Broth Type Tryptic Soy Broth
Manufacturing Origin United States
Product Type Broth Tube
Sterility Non-sterile
Target Organism Class Bacteria, Fungi
Test Type Microbial Detection, Growth Promotion Testing
Packaging Type Glass Tube
Quantity 15 mL
Dimensions (Dia. x H) 20 x 125 mm