missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptic Soy Broth

Catalog No. R117834
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
16mm x 125mm Tube
16mm x 125mm Tube, Hungate Cap
Tube
Fill Volume:
10 mL HC
9 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Fill Volume
R117834 20/Pk. 16mm x 125mm Tube, Hungate Cap 10 mL HC
R07226 100/Pk. Tube 10 mL
R117832 100/Pk. 16mm x 125mm Tube 9 mL
R07228 20/Pk. Tube 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R117834 Supplier Thermo Scientific™ Supplier No. R117834

Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.

Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use TSB tubes.

Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Description Tryptic Soy Broth
Format 16mm x 125mm Tube, Hungate Cap
Manufacturing Origin United States
Product Type Microbiology Media
Fill Volume 10 mL HC
Quantity 20/Pk.