missing translation for 'onlineSavingsMsg'
Learn More

Hardy Diagnostics™ Tryptic Soy Agar

Catalog No. 23001759
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23001759 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23001759 Supplier Hardy Diagnostics™ Supplier No. G60BX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use as a non-selective, general growth medium for the isolation and cultivation of microorganisms (bacteria, yeast, and fungi)

  • Especially useful for performing colony counts
  • This medium is also recommended for use in the cultivation, storage, maintenance, and transportation of pure cultures of microorganisms
  • Optically clear polystyrene for distortion free viewing

Specifications

Appearance Translucent
Certifications/Compliance ISO 17025
Color Light Amber
Content And Storage 2°C to 8°C
Country of Origin United States
Detects Microorganisms (Bacteria, Yeast, and Fungi)
Form Prepared Medium
Format Plate
Manufacturing Origin United States
Product Type Agar Plate
Sterility Sterile
Target Organism Class Yeast, Bacteria, Fungi
Test Type Standard Plate Count for Microorganism
Fill Volume 18 mL
Formulation Pancreatic Digest of Casein - 15.0 g/L, Peptic Digest of Soybean Meal - 5.0 g/L, Sodium Chloride - 5.0 g/L, Agar - 15.0 g/L, Dipotassium Phosphate - 2.5 g/L
For Use With (Application) Isolation and cultivation of microorganisms (bacteria, yeast, and fungi
Incubation Conditions 30°C to 35°C, 20°C to 25°C
Material Polystyrene
No. per Pack 100
Packaging Type Sleeve
Quantity 100 Plates
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Staphylococcus species, Escherichia species, Pseudomonas species, Bacillus species, Salmonella species
Protein Source Casein, Soybean Meal
Manufacturing Quality ISO 17025
Show More Show Less