missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine 98.0+%, TCI America™

Catalog No. T089025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T089025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T089025G Supplier TCI America Supplier No. T089025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61-54-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00005661
InChI Key APJYDQYYACXCRM-UHFFFAOYSA-N
Synonym tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
PubChem CID 1150
ChEBI CHEBI:16765
IUPAC Name 2-(1H-indol-3-yl)ethan-1-amine
SMILES NCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 117°C
Quantity 25 g
Formula Weight 160.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tryptamine
EINECSNumber (5)-5472
RTECSNumber NL4020000
TSCA No
Recommended Storage Refrigerator