missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine Hydrochloride 98.0+%, TCI America™

Catalog No. A03001G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03001G 1 g
A030025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03001G Supplier TCI America Supplier No. A03001G

Chemical Identifiers

CAS 343-94-2
Molecular Formula C10H13ClN2
Molecular Weight (g/mol) 196.678
MDL Number MFCD00012682
InChI Key KDFBGNBTTMPNIG-UHFFFAOYSA-N
Synonym tryptamine hydrochloride, 3-2-aminoethyl indole hydrochloride, 2-1h-indol-3-yl ethanamine hydrochloride, 1h-indole-3-ethanamine, monohydrochloride, tryptamine monohydrochloride, tryptaminehydrochloride, ccris 4419, indole-3-ethylamine hydrochloride, 3-2-aminoethyl-1h-indole monohydrochloride, beta-indole-ethylamine hydrochloride
PubChem CID 67652
IUPAC Name 2-(1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)C(=CN2)CCN.Cl

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tryptamine Hydrochloride
RTECSNumber NL4375000
TSCA Yes