missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine hydrochloride, 98%

Catalog No. AC157980050
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157980050 5 g Glass bottle
AC157980100 10 g Glass bottle
AC157981000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157980050 Supplier Thermo Scientific Chemicals Supplier No. 157980050

CAS: 343-94-2 | C10H12N2·HCl | 196.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 343-94-2
Molecular Formula C10H12N2·HCl
Molecular Weight (g/mol) 196.68
MDL Number MFCD00012682
InChI Key KDFBGNBTTMPNIG-UHFFFAOYSA-N
Synonym tryptamine hydrochloride, 3-2-aminoethyl indole hydrochloride, 2-1h-indol-3-yl ethanamine hydrochloride, 1h-indole-3-ethanamine, monohydrochloride, tryptamine monohydrochloride, tryptaminehydrochloride, ccris 4419, indole-3-ethylamine hydrochloride, 3-2-aminoethyl-1h-indole monohydrochloride, beta-indole-ethylamine hydrochloride
PubChem CID 67652
IUPAC Name 2-(1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)C(=CN2)CCN.Cl

Specifications

Melting Point 253°C to 255°C
Color Beige to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 22, I, 636
Merck Index 14, 9796
Solubility Information Solubility in water: 64g/L (20°C)
Formula Weight 196.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tryptamine hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-446-4
RTECSNumber NL4375000
TSCA TSCA

RUO – Research Use Only