missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trypsin Inhibitor, soybeans

Catalog No. AAJ6098203
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6098203 1 g
AAJ60982MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6098203 Supplier Thermo Scientific Chemicals Supplier No. J6098203

Inhibits the pancreatic enzyme trypsin | CAS: 9035-81-8

The soybean trypsin inhibitor has been tested for cell culture applications. It has been used in a study to assess the potential application in animal cell culture of an alkaline protease from a non-toxigenic mangrove isolate of Vibri sp. V26. It has also been used in a study to improve the detection of fungi in eosinophilic mucin. It is an inhibitor of trypsin factor Xa, plasmin and neutrophil elastase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Dissolve in 1X PBS at 1 to 10mg/ml for stock solution

Chemical Identifiers

CAS 9035-81-8

Specifications

Quantity 1 g
Description Off-white to Pale Brown
Content And Storage 2°C to 8°C
For Use With (Application) Inhibits the pancreatic enzyme trypsin
Product Type Signal Transduction Reagent
Form Powder
EINECSNumber 232-906-9
Recommended Storage Keep cold

RUO – Research Use Only