missing translation for 'onlineSavingsMsg'
Learn More

Trypsin inhibitor, MP Biomedicals™

Catalog No. ICN10111325
Change view
Click to view available options
Quantity:
25 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10111325 25 mg
ICN10111394 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10111325 Supplier MP Biomedicals Inc Supplier No. 0210111325

Chemical Identifiers

CAS 9035-81-8
Synonym Ovomucoid trypsin inhibitor

Specifications

Color White
Quantity 25 mg
Formula Weight 28,500 ± 3,5003,4,5
Physical Form Powder
Chemical Name or Material Trypsin Inhibitor