missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypan Blue Stain Solution, MP Biomedicals
0.4% (w/v) Solution In normal saline Storage temperature 15-30°C
Supplier: MP Biomedicals Inc 091691049
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Trypan Blue Stain Solution | |
C34H24N6Na4O14S4 | |
WMYVHJWZUUEZNE-ARWFNKCKSA-J | |
tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate | |
9562061 |
72-57-1 | |
MFCD00003969 | |
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 | |
960.79 | |
100 mL |
Chemical Identifiers
72-57-1 | |
960.79 | |
WMYVHJWZUUEZNE-ARWFNKCKSA-J | |
tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonateShow More |
C34H24N6Na4O14S4 | |
MFCD00003969 | |
9562061 | |
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1Show More |