Learn More
Thermo Scientific Chemicals Trypan Blue
CAS: 72-57-1 | C34H24N6Na4O14S4 | 960.79 g/mol
Supplier: Thermo Scientific Chemicals 189351000
| Quantity | 100 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C34H24N6Na4O14S4 | |
| MFCD00003969 | |
| C.I. 23850, Direct Blue 14, Niagara Blue 3B | |
| [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 |
Specifications
| Trypan Blue | |
| 72-57-1 | |
| C34H24N6Na4O14S4 | |
| C.I. 23850, Direct Blue 14, Niagara Blue 3B | |
| WMYVHJWZUUEZNE-ARWFNKCKSA-J | |
| Authentic | |
| 960.79 | |
| 960.78 | |
| Complies | |
| 100 g |
| >280.0°C | |
| 60% min. | |
| MFCD00003969 | |
| Solubility in water: 10g/L (25°C). Other solubilities: practically insoluble in alcohol, soluble in cellosolve, ethylene glycol, acids | |
| [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 | |
| tetrasodium;(3Z)-5-amino-3-[[4-[4-[(2Z)-2-(8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate | |
| 9562061 | |
| Glass bottle | |
| Black/Brown to Blue | |
| Powder |
Safety and Handling
GHS P Statement
May cause cancer.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 200-786-7
RTECSNumber : QJ6475000
TSCA : TSCA