missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trypan Blue

Catalog No. AC189350250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189350250 25 g Glass bottle
AC189351000 100 g Glass bottle
AC189355000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC189350250 Supplier Thermo Scientific Chemicals Supplier No. 189350250

CAS: 72-57-1 | C34H24N6Na4O14S4 | 960.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72-57-1
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00003969
InChI Key WMYVHJWZUUEZNE-ARWFNKCKSA-J
Synonym C.I. 23850, Direct Blue 14, Niagara Blue 3B
PubChem CID 9562061
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Melting Point >280.0°C
Dye Content 60% min.
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: practically insoluble in alcohol, soluble in cellosolve, ethylene glycol, acids
Infrared Spectrum Authentic
IUPAC Name tetrasodium;(3Z)-5-amino-3-[[4-[4-[(2Z)-2-(8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
Formula Weight 960.78
Packaging Glass bottle
UV Spectrometry Complies
Color Black/Brown to Blue
Quantity 25 g
Physical Form Powder
Chemical Name or Material Trypan Blue
Show More Show Less
Hazard Statement GHS P Statement
May cause cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-786-7
RTECSNumber QJ6475000
TSCA TSCA

RUO – Research Use Only