Learn More
Stemcell Technologies Inc Trypan Blue, Size: 100 mL
Catalog No. NC9996409
Encompass
Trypan Blue is recommended for counting viable mammalian cells. Viable cell counts should be performed where a decrease in cell viability may be expected, for example, when working with cryopreserved cells or cells manipulated ex vivo.
Catalog No. NC9996409 Supplier Stemcell Technologies Inc Supplier No. 07050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Trypan Blue is recommended for counting viable mammalian cells. Viable cell counts should be performed where a decrease in cell viability may be expected, for example, when working with cryopreserved cells or cells manipulated ex vivo.