Learn More
Trundle Wheel
Measures long distances in the classroom or outdoors.
Supplier: Customer Channels Group (Ccg) 45550
Description
- Weather-resistant, all-plastic construction
- Loud, non-wearing clicking device is activated in either direction with each full meter revolution
- Comes apart in two pieces for storage and is easily reassembled
- Counter measures up to 99,999m with adjustable handle
- Tire is equipped with a rubber tread to provide accurate readings