missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Trundle Wheel

Catalog No. S15831
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S15831 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15831 Supplier United Scientific™ Supplier No. TRUNWHL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly durable compared to other versions on the market.

  • This trundle wheel easily rolls along the ground to measure linear distance.
  • A metered ounter measures up to 99,999 meters, and the rubber wheel edge provides a steady rip when rolling over different surfaces.
  • Telescoping handle can extend from 43 cm to 6 cm to provide comfortable operation for all users.
  • The built-in folding stand allows pright storage.
  • Includes a convenient carrying case.Includes storage case
  • Displays in increments of 0.2 m
  • Wheel has 1 cm markings along the edge

Specifications

Grade 5 to 12
Includes Trundle Wheel and Storage Case
Class Size 1 Lab Group
Product Type Trundle Wheel

Warning: Cancer and reproductive harm