missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ TRUFLOW™ Tissue and Biopsy Cassettes

Catalog No. 15200404E
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Product Type:
Biopsy Cassette
Tissue Cassette
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
15200404E White Biopsy Cassette
15200403D Blue Tissue Cassette
15200403C Green Tissue Cassette
15200403A Yellow Tissue Cassette
15200403B Pink Tissue Cassette
15200403E White Tissue Cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15200404E

Just bend, snap apart, and conveniently lock

  • Hold tissue and biopsy specimens during the embedding process as well as in a storage file
  • Molded from a special high-density polymer that is totally resistant to histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Can be opened or closed as often as necessary and relocked securely without danger of specimen loss
  • Large labeling areas are located on three sides of the cassette
  • Anterior writing area at a 30° angle

Order Info

Packaged in three dispenser boxes of 500

Specifications

Pore Style Slots
Writing Face Angle 30°
Closure Type Integral Lid
Color White
Material High Density Polymer
Product Type Biopsy Cassette