missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueGuide™ sgRNA Negative Control, non-targeting 1

Catalog No. A35526
Encompass
Change view
Click to view available options
Quantity:
3 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35526 3 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A35526 Supplier Invitrogen™ Supplier No. A35526

missing translation for 'validMainframeMsg'

TrueGuide Synthetic sgRNA Negative Control does not recognize any sequence in the human or mouse genome and provides a non-edited experimental control.

TrueGuide negative controls are non-targeting gRNA sequences that do not recognize any sequence in the human or mouse genome and provide a non-edited control when performing analysis on pooled populations of edited cells.

Non-targeting gRNA sequences that do not recognize any sequence in the human or mouse genome and provide a non-edited control when performing analysis on pooled populations of edited cells.

Validated negative control crRNA. Universal control for Mouse and Human

Specifications

Content And Storage Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5°C to -30°C. Resuspend in nuclease-free water or TE.
Format Tube
Control Type Negative Control
RNAi Type gRNA
Species Human, Mouse
Product Line TrueGuide
Product Type gRNA
Quantity 3 nmol
Shipping Condition Room Temperature
I would like to calculate the molecular weight of TrueGuide Synthetic gRNA? What is the length of TrueGuide Synthetic gRNA (TrueGuide sgRNA)?

The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.

For Research Use Only. Not for use in diagnostic procedures.