Learn More
Perfex TruCLEAN Pro Triple-Bucket
Catalog No. p-200003930
Change view
Click to view available options
Color:
Green
White
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19005390 White
19005392 Green
19005393 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19005390 Supplier Perfex Supplier No. 223W
Only null left
Heavy-duty stainless-steel cart with multiple configurations, colorways, and accessories to customize for your specific environment.
Capture and isolate contaminants to ensure the delivery of unadulterated cleaning and sanitizing agents. No more re-applying dirty water and weakened solutions. TruCLEAN Pro systems efficiently clean and disinfect floors, walls, and ceilings in large or small area cleanrooms of any grade.
The adaptable TruCLEAN Pro system allows triple (15 L) or double (22 L) bucket configuration in a heavy-duty stainless-steel cart designed for strength, durability, and hygiene. Our Pro series offers the most versatile mopping equipment with multiple configurations, colorways, and accessories to customize for your specific environment.
Each mopping system component plays an important role in delivering the highest level of cleanliness. For example, our innovative swivel mop frame is accurately balanced to maintain total surface contact at any angle. The unique wringing mechanism exerts force evenly across the entire mop surface, extracting the maximum amount of soiled contaminants.
Specifications
Color White
Dimensions (L x W x H) 27 x 19 x 41 in.