Learn More
Perfex TruCLEAN Disinfection
Quickly apply disinfecting agents onto floors, walls, and ceilings. Compact design for simplified use in confined areas.
Supplier:  Perfex 301G
 View more versions of this product
Catalog No. 19005375
Only null left
Description
Description
Designed for quick application of disinfecting agents onto floors, walls, and ceilings. Simply dip the mop in solution, press mop on dosage sieve and apply to surface. Compact design is perfect for use in confined areas, allows easy storage with limited space.
Temperature-resistant 36 Liter polypropylene bucket includes embossed graduations for accurate measure and 3-inch twin-wheel casters with one locking caster. Easily accessible brake pedal allows parking on uneven floors. Stainless steel dosage sieve mounts evenly while precisely positioned apertures release excess disinfectant into 36 Liter bucket.
Product Suggestions
Product Suggestions