missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropylium Tetrafluoroborate 98.0+%, TCI America™
Supplier: TCI America T11215G
Specifications
| Tropylium Tetrafluoroborate | |
| C7H7BF4 | |
| 5 g | |
| tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t | |
| F[B-](F)(F)F.[CH+]1C=CC=CC=C1 | |
| 177.94 | |
| 177.94 | |
| Crystalline Powder |
| 27081-10-3 | |
| MFCD00013402 | |
| 1759 | |
| SQVQHTIKOZVGLR-UHFFFAOYSA-N | |
| cyclohepta-2,4,6-trien-1-ylium; tetrafluoroboranuide | |
| 152926 | |
| ≥98.0% (T) |
Chemical Identifiers
| 27081-10-3 | |
| 177.94 | |
| SQVQHTIKOZVGLR-UHFFFAOYSA-N | |
| 152926 | |
| F[B-](F)(F)F.[CH+]1C=CC=CC=C1 |
| C7H7BF4 | |
| MFCD00013402 | |
| tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t | |
| cyclohepta-2,4,6-trien-1-ylium; tetrafluoroboranuide |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator